Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sanfetrinem sodium | 141611-76-9 | 95.0% | 303.29 g·mol⁻1 | C14H18NNaO5 | 1 MG
SDP

Catalog No. 5000239805
Encompass_Preferred

Sanfetrinem sodium is the sodium salt of sanfetrinem, a beta-lactamase-stable, broad-spectrum antibiotic supplied as a solid research compound for in vitro and related studies.

  • Provides beta-lactamase-stable broad-spectrum antibacterial activity.
  • CAS number 141611-76-9.
  • Molecular formula C14H18NNaO5; molecular weight 303.29 g·mol⁻1.
  • Supplied as a solid sodium salt suitable for laboratory research.
  • Typical purity (NMR) 95.0% as reported on the certificate of analysis.
  • Available in small pack sizes for screening and analytical use.

Catalog No. 50-002-39805 Supplier Medchemexpress LLC Supplier No. HY106922A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sanfetrinem sodium is the sodium salt of sanfetrinem, a beta-lactamase-stable, broad-spectrum antibiotic supplied as a solid research compound for in vitro and related studies.

  • Provides beta-lactamase-stable broad-spectrum antibacterial activity.
  • CAS number 141611-76-9.
  • Molecular formula C14H18NNaO5; molecular weight 303.29 g·mol⁻1.
  • Supplied as a solid sodium salt suitable for laboratory research.
  • Typical purity (NMR) 95.0% as reported on the certificate of analysis.
  • Available in small pack sizes for screening and analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.