Learn More
TARGETMOL CHEMICALS INC SANGGENON C 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4S16155MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Sanggenon C (Sanggenone C) inhibits tumor cellular proteasomal activity and cell viability via induction of cell cycle arrest and cell death inhibiting the proteasome function. 2. Sanggenon C and O inhibited NO production and iNOS expression by suppressing NF-(kappa)B activity and I(kappa)B(alpha) activation; inhibited TNF-alpha -stimulated PMN-HSC adhesion and expression of VCAM-1 by suppressing the activation of NF-kappaB. purity: 98%
Catalog No. 50-257-9831