Learn More
TARGETMOL CHEMICALS INC Sanggenon D 1MG
SDP
Catalog No. 502876734
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes.Sanggenon D (Sanggenone D), a small molecule compound from Morus alba Bark, is a GABA(A) receptor modulator with antioxidant and antibacterial activity, inhibits breast cancer cell proliferation, and inhibits the growth of Staphylococcus aureus by modulating the fatty acid biosynthesis system. Purity 98%
Catalog No. 50-287-6734 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S16161MG
Also available in 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes.Sanggenon D (Sanggenone D), a small molecule compound from Morus alba Bark, is a GABA(A) receptor modulator with antioxidant and antibacterial activity, inhibits breast cancer cell proliferation, and inhibits the growth of Staphylococcus aureus by modulating the fatty acid biosynthesis system. Purity 98%