Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sangivamycin | 18417-89-5 | 97.7% | 309.28 g/mol | C12H15N5O5 | 1 MG
SDP

Catalog No. 5000307666
Encompass_Preferred

Sangivamycin is a nucleoside analog research reagent with protein kinase C (PKC) inhibitory activity and nucleoside antimetabolite properties. It is supplied for in vitro and in vivo research, with recommended solubility and storage conditions for experimental use.

  • Nucleoside analog with PKC inhibitory activity.
  • CAS number 18417-89-5 for unambiguous identification.
  • Molecular formula C12H15N5O5; molecular weight 309.28 g/mol.
  • High reported purity (97.68%).
  • DMSO solubility: 100 mg/mL; in vivo formulations available (solubility ≥ 2.5 mg/mL).
  • Storage recommendations: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-07666 Supplier Medchemexpress LLC Supplier No. HY1183841MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sangivamycin is a nucleoside analog research reagent with protein kinase C (PKC) inhibitory activity and nucleoside antimetabolite properties. It is supplied for in vitro and in vivo research, with recommended solubility and storage conditions for experimental use.

  • Nucleoside analog with PKC inhibitory activity.
  • CAS number 18417-89-5 for unambiguous identification.
  • Molecular formula C12H15N5O5; molecular weight 309.28 g/mol.
  • High reported purity (97.68%).
  • DMSO solubility: 100 mg/mL; in vivo formulations available (solubility ≥ 2.5 mg/mL).
  • Storage recommendations: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.