Learn More
Selleck Chemical LLC Sanguinarine-1mg
SDP
Supplier:  Selleck Chemical LLC S90321MG
Encompass_Preferred
Sanguinarine (Pseudochelerythrine, Sanguinarin, Sanguinarium), a benzophenanthridine alkaloid known as an anti-inflammatory agent, is a potent inhibitor of NF-κB activation.
Catalog No. 50-238-9800