Learn More
Medchemexpress LLC Sanguinarine chloride | 5578-73-4 | MFCD00012126 | 98.7% | 367.78 g/mol | C20H14ClNO4 | 10 MG
SDP
Catalog No. 5000299000
Encompass_Preferred
Sanguinarine chloride is a benzophenanthridine alkaloid supplied as a research-grade analytical standard for biochemical and cell biology studies, notably apoptosis and autophagy assays. It is provided as a small, high-purity reagent intended for analytical and laboratory use.
  • High purity (98.7%) suitable for analytical use.
  • Molecular weight 367.78 g/mol and chemical formula C20H14ClNO4.
  • Identified by CAS number 5578-73-4 for unambiguous reference.
  • Provided as a 10 MG pack for laboratory use.
  • Suitable as a standard in cell biology and biochemical assays.
Catalog No. 50-002-99000 Supplier Medchemexpress LLC Supplier No. HYN0052A10MG
Add to Cart
Add to Cart
Sanguinarine chloride is a benzophenanthridine alkaloid supplied as a research-grade analytical standard for biochemical and cell biology studies, notably apoptosis and autophagy assays. It is provided as a small, high-purity reagent intended for analytical and laboratory use.
  • High purity (98.7%) suitable for analytical use.
  • Molecular weight 367.78 g/mol and chemical formula C20H14ClNO4.
  • Identified by CAS number 5578-73-4 for unambiguous reference.
  • Provided as a 10 MG pack for laboratory use.
  • Suitable as a standard in cell biology and biochemical assays.