Learn More
Enzo Life Sciences Sanguinarine chloride (5mg). CAS: 5578-73-4
SDP
Supplier:  Enzo Life Sciences ALX350076M005
Encompass_Preferred
Na+/K+-and Mg2+-ATPase inhibitor. Antimicrobial, anti-inflammatory and antioxidant properties. Induces apoptosis. Modulates the Bcl-2 family. Shows antiplaque activity in humans. Alternative name: Pseudochelerythrine. Purity: ≥98% (HPLC). Appearance: Orange solid. Solubility: Soluble in methanol; slightly soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-8719