Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sanguinarine chloride, Thermo Scientific™

Catalog No. AC468962500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468962500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC468962500 Supplier Thermo Scientific Chemicals Supplier No. 468962500
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Light Yellow to Dark Orange
Description Sanguinarine chloride
CAS 5578-73-4
Molecular Formula C20 H14 Cl N O4
Shipping Condition No special requirements
Product Line Thermo Scientific
Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.