Learn More
Biosan Laboratories Sani-Check AB Kit, 25 Tests
Catalog No. NC9188280
Encompass_Preferred
Bacterial growth in the industrial environment can often lead to serious economic consequences. Elevated bacterial populations in aqueous systems such as cooling waters, metalworking fluids, and hydrocarbon fuels are responsible for extensive losses in productivity. Problems related to high levels of bacteria in these liquids include corrosion, slime formation, foul odors, and general decreases in fluid function. Effective biocide treatment is possible only if the degree of of bacterial contamination is routinely determined. Sani-Check AB makes it fast and easy to monitor aerobic bacterial count.
Catalog No. NC9188280 Supplier Biosan Laboratories Supplier No. 110
Bacterial growth in the industrial environment can often lead to serious economic consequences. Elevated bacterial populations in aqueous systems such as cooling waters, metalworking fluids, and hydrocarbon fuels are responsible for extensive losses in productivity. Problems related to high levels of bacteria in these liquids include corrosion, slime formation, foul odors, and general decreases in fluid function. Effective biocide treatment is possible only if the degree of of bacterial contamination is routinely determined. Sani-Check AB makes it fast and easy to monitor aerobic bacterial count.