Learn More
Biosan Laboratories SANI-CHECK BACTERIA COUNT/10PK
Catalog No. NC9570593
Encompass_Preferred
Sani-Check B, Test System for Counting Bacteria, Six Month Shelf, Stored in the Refrigerator or at Room Temperature, Contain Paddles Coated with Agar Culture Medium, Makes Determination of Microbial Counts Fast and Easy
Catalog No. NC9570593 Supplier Biosan Laboratories Supplier No. 210
Sani-Check B, Test System for Counting Bacteria, Six Month Shelf, Stored in the Refrigerator or at Room Temperature, Contain Paddles Coated with Agar Culture Medium, Makes Determination of Microbial Counts Fast and Easy