Learn More
Sentinel Process Systems SANI CLAMP GSK 1/2IN SPC
SDP
Catalog No. NC1700299
Encompass
Sanitary Clamp GSK 1/2in SIL PLTNM CURED - BioPure USP Class VI; FDA Compliant; Platinum Cured Clear Silicone Sanitary Clamp Gasket; 1/2in.
Catalog No. NC1700299 Supplier Sentinel Process Systems Supplier No. 42RXPX050
Sanitary Clamp GSK 1/2in SIL PLTNM CURED - BioPure USP Class VI; FDA Compliant; Platinum Cured Clear Silicone Sanitary Clamp Gasket; 1/2in.