Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Sani/Clorox Wipe Holder
SDP

Catalog No. 22000387
Change view
Click to view available options
Mounting:
Surface Mount
Wall Mount
No. of Compartments:
1
2
3
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting No. of Compartments
22-000-387 Wall Mount 3
22-000-382 Surface Mount 1
22-000-383 Wall Mount 1
22-000-384 Surface Mount 2
22-000-385 Wall Mount 2
22-000-386 Surface Mount 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-000-387 Supplier Poltex Supplier No. SANICLOR3W
  • $178.00
    $127.65 / Each of 1

    Save $50.35

    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Holds wipe tubs.

Specifications

Product Type Wipe Tub Holder
Color Clear
Length (Metric) 147 mm
Width (Metric) 406 mm
Height (Metric) 228 mm
Material PETG
Mounting Wall Mount
No. of Compartments 3
For Use With (Application) Sani or clorox wipes or equivalent
Quantity 1 Ea.
Thickness (English) 0.25 in.
Show More Show Less

Consumables not included.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.