Learn More
Med Vet International Dynarex Sani-Cloth Plus, L, 1920/Case
Supplier:  Med Vet International PQ89072
Encompass_Preferred
Effective against 16 microorganisms in 3 minutes. Ideal for use in alternate care settings including physician and dental offices. Contains 160 wipes Multi-count canister accommodates frequent usage EPA-registered low level disinfectant Pre-moistened wipes are ready to use16 microorganisms Compares to competitor Quat/Low-Alcohol disinfectant products Compatible with a broad range of surfaces and equipment in healthcare Meets CDC, OSHA and infection control guidelines Deep well lid provides ample space to store and access next wipe Prominent contact time for easy identification and improved compliance with accrediation standards Categorization of pathogens allows for quick identification of relevant efficacy claims Icon clearly communicates key safety information on both label and lid Point of care accessories and tools are available to help improve staff compliance to facility protocols One step cleaner and disinfectant May be used on hard nonporous surfaces.
Catalog No. 50-282-9405