Learn More
Med Vet International Dynarex Sani-Cloth Super, L, 1920/Case
Catalog No. 502829803
Encompass_Preferred
Effective against 30 microorganisms in 2 minutes. Fast contact time allows for a quick room turnover. Ideal for daily use in fast-paced environments that require short contact times and broad coverage of microorganisms. Contains 160 wipes Ulti-count canister accommodates frequent usage EPA-registered intermediate level disinfectant Thick, strong wipe has high fluid capacity and is pliable to get into crevices and corners of equipment Pre-moistened wipes are ready to use30 microorganisms,  including 6 multi-drug resistant organisms (MDROs)Compatible with a broad range of surfaces and equipment in healthcare Meets CDC, OSHA and CMS Tag F441 guidelines Deep well lid provides ample space to store and access next wipe Prominent contact time for easy identification and improved compliance with accrediation standards
Catalog No. 50-282-9803 Supplier Med Vet International Supplier No. PQ55172
 Restricted Item
Add to Cart
Add to Cart
Effective against 30 microorganisms in 2 minutes. Fast contact time allows for a quick room turnover. Ideal for daily use in fast-paced environments that require short contact times and broad coverage of microorganisms. Contains 160 wipes Ulti-count canister accommodates frequent usage EPA-registered intermediate level disinfectant Thick, strong wipe has high fluid capacity and is pliable to get into crevices and corners of equipment Pre-moistened wipes are ready to use30 microorganisms,  including 6 multi-drug resistant organisms (MDROs)Compatible with a broad range of surfaces and equipment in healthcare Meets CDC, OSHA and CMS Tag F441 guidelines Deep well lid provides ample space to store and access next wipe Prominent contact time for easy identification and improved compliance with accrediation standards