Learn More
Med Vet International Dynarex Sani-Cloth Super, Xl, 390/Case
Supplier:  Med Vet International PQ86984
Encompass_Preferred
The number one disinfecting wipe in healthcare.  Effective against 30 microorganisms in 2 minutes. Fast contact time allows for a quick room turnover. Ideal for daily use in fast-paced environments that require short contact times and broad coverage of microorganisms. Contains 65 x-large wipes Perfect wipe size for larger surface areas Multi-count canister accommodates frequent usage EPA-registered intermediate level disinfectant Thick, strong wipe has high fluid capacity and is pliable to get into crevices and corners of equipment Pre-moistened wipes are ready to use30 microorganisms,  including 6 multi-drug resistant organisms (MDROs)Compatible with a broad range of surfaces and equipment in healthcare Meets CDC, OSHA and CMS Tag F441 guidelines Deep well lid provides ample space to store and access next wipe Prominent contact time for easy identification and improved compliance with accrediation standards.
Catalog No. 50-283-1791