Learn More
McMaster-Carr SANI QUICK-CLAMP FIT TEFL 3/4
Catalog No. NC1306182
Encompass
Gasket for Sanitary Quick-Clamp Fitting Clean Room Teflon, for 3/4" Tube OD
Catalog No. NC1306182 Supplier McMaster-Carr Supplier No. 1314N42
Gasket for Sanitary Quick-Clamp Fitting Clean Room Teflon, for 3/4" Tube OD