Learn More
World Precision Instrument Sani-Safe Autopsy grossing Knives, 9in
SDP
Catalog No. 502538276
Encompass
Sani-Safe Autopsy Grossing Knife is crafted from premium grade stainless steel, which may be repeated sharpened, as needed. Its knurled handle is designed for comfortable and secure use, and it is solidly mounted for safety in use. This sharp, drop point knife is formed for gross dissection, giving you full control of the cutting edge and years of service.
Catalog No. 50-253-8276 Supplier World Precision Instrument Supplier No. WP9111
Sani-Safe Autopsy Grossing Knife is crafted from premium grade stainless steel, which may be repeated sharpened, as needed. Its knurled handle is designed for comfortable and secure use, and it is solidly mounted for safety in use. This sharp, drop point knife is formed for gross dissection, giving you full control of the cutting edge and years of service.