Learn More
BEST SANITIZERS Alpet E3 Plus Hand Sanitizer Spray, 1 Liter
Catalog No. NC1123418
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Alpet E3 Plus Hand Sanitizer Spray is an atomized alcohol hand sanitizing formula engineered to meet the requirements of the Food Industry. Alpet E3 Plus meets all current FDA and USDA requirements for hand sanitizers in food handling environments. Alpet E3 Plus is a powerful, yet gentle ethanol-based formula which has been tested and proven effective against 26 pathogens. Kills 99.9999% (LOG6) of 26 tested pathogens in 15 seconds Atomized spray provides better coverage Meets all FDA (GRAS) and USDA requirements for food handling Formulated with moisturizers to keep hands soft and healthy NSF certified E3 classification Kosher, Pareve and Halal certified 71% Ethanol formula
Catalog No. NC1123418 Supplier BEST SANITIZERS Supplier No. SA10014
This item has a minimum qty of 2 per supplier requirements.
Alpet E3 Plus Hand Sanitizer Spray is an atomized alcohol hand sanitizing formula engineered to meet the requirements of the Food Industry. Alpet E3 Plus meets all current FDA and USDA requirements for hand sanitizers in food handling environments. Alpet E3 Plus is a powerful, yet gentle ethanol-based formula which has been tested and proven effective against 26 pathogens. Kills 99.9999% (LOG6) of 26 tested pathogens in 15 seconds Atomized spray provides better coverage Meets all FDA (GRAS) and USDA requirements for food handling Formulated with moisturizers to keep hands soft and healthy NSF certified E3 classification Kosher, Pareve and Halal certified 71% Ethanol formula