Learn More
Accuflow Systems Sani-Tech® SPT-60L
Catalog No. NC3955483
Encompass_Preferred
Sani-Tech® SPT-60L is premium platinum-cured silicone tubing engineered for demanding pharmaceutical operations. It offers best-in-class accuracy, precision, and minimal spallation, ensuring consistent filling performance and extended pump life.
  • Long pump life
  • Low spallation
  • Accurate and consistent dispensing
  • Full extractables report per BioPhorum Operations Group (BPOG) protocol
  • Technical dossier available
  • Available with ValPlus™ certification
Catalog No. NC3955483 Supplier Accuflow Systems Supplier No. SPT60L047157
Sani-Tech® SPT-60L is premium platinum-cured silicone tubing engineered for demanding pharmaceutical operations. It offers best-in-class accuracy, precision, and minimal spallation, ensuring consistent filling performance and extended pump life.
  • Long pump life
  • Low spallation
  • Accurate and consistent dispensing
  • Full extractables report per BioPhorum Operations Group (BPOG) protocol
  • Technical dossier available
  • Available with ValPlus™ certification