Learn More
Mopec SaniPath Cleaning and Disinfecting Wipes
SDP
Catalog No. 22444300 Shop All Mopec Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
22-444-300 Clear
1 options
This item is not returnable. View return policy
Catalog No. 22-444-300 Supplier Mopec Supplier No. BE036
  • $24.75
    $22.65 / Pack of 160
    Save $2.10
    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mopec SaniPath Cleaning and Disinfecting Wipes offer one-step cleaning and disinfecting for all pathology/necropsy lab surfaces
  • Allow you to clean and disinfect in a pinch with or without gloves
  • Effective against a wide range of pathogenic bacteria, viruses and fungi such as HIV-1, H1N1, MRSA, Herpes Simplex 2, Pseudomonas, Staphylococcus, Tuberculosis and Canine Parvo
  • Ideal for lab telephones, computers, keyboards, writing instruments and splash shields.
Specifications
Product Type Disinfectant Wipe
Material Substrate
Color Clear
Description Disinfecting Wipes
Cannot ship to California.