Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec SaniPath Disinfectant Foaming Spray
SDP

Catalog No. 22444304 Shop All Mopec Products
Change view
Click to view available options
pH:
11.5 to 12.5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
22-444-304 11.5 to 12.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-444-304 Supplier Mopec Supplier No. BE045
  • $44.50
    $37.65 / Each of 1

    Save $6.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mopec SaniPath Disinfectant Foaming Spray is a powerful foaming spray that destroys a wide range of bacteria, viruses and fungi

  • Multi-purpose decontaminating cleaner
  • Bactericidal, fungicidal, virucidal and Tuberculocide
  • Destroys a wide range of pathogenic bacteria, viruses and fungi including HIV-1, HIN1, Herpes Simplex 2, Staphylococcus, Pseudomonas, Canine Parvo and Tuberculosis
  • Clean, disinfect and deodorize in 10 minutes with the clinging foam
  • Mild fragrance leaves your lab smelling fresh
  • Meets OSHAs Bloodboorne Pathogens Standard for decontamination of hard, non-porous environmental surfaces
  • Not available in California

Specifications

Product Type Disinfectant
Color-coded No
Form Liquid Spray
pH 11.5 to 12.5
Scent Unscented
Sterility Sterile
Appearance Colorless
Container Type Spray Bottle
Content And Storage Store between 50 and 120°F
Description Disinfectant Foaming Spray

Cannot ship to California.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.