Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec SaniPath Disinfecting Spray Cleaner
SDP

Catalog No. 22444306 Shop All Mopec Products
Change view
Click to view available options
pH:
11.5 to 12.5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
22-444-306 11.5 to 12.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-444-306 Supplier Mopec Supplier No. BE047
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mopec SaniPath Disinfecting Spray Cleaner is bactericidal, fungicidal, virucidal and tuberculocide

  • Multi-purpose decontaminating cleaner
  • Mild, lemon fragrance leaves your lab smelling fresh
  • Meets OSHAs Bloodboorne Pathogens Standard for decontamination of hard, non-porous environmental surfaces
  • Not available in California

Specifications

Product Type Disinfectant
Color-coded No
Form Spray
pH 11.5 to 12.5
Scent Unscented
Sterility Sterile
Appearance Blue
Container Type Spray Bottle
Content And Storage Store between 50 and 120°F
Description Disinfecting Spray

Cannot ship to California.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.