Learn More
Sanisure Inc Bio-Ease+™ Sanitary Clamps .75"
Supplier:  Sanisure Inc 13MHHS07501 EA
Encompass_Preferred
SaniSure Bio-Ease+™ is a patented non-metallic sanitary clamp designed to meet the needs of various single-use applications in the pharmaceutical and bio-processing industries. Manufactured using a glass-reinforced, USP Class VI Nylon and designed to be used as a component for single-use transfer lines and a wide range of bio-process systems, these plastic clamps are much lighter than their stainless steel counterparts. SaniSure stocks a comprehensive inventory of sizes from Mini to 4″. Additionally, these clamps fit ASME BPE Tri-Clamp connections, ensuring compatibility with industry-standard equipment.
Catalog No. NC3662604