Learn More
Cole-Parmer SANITARY CLAMP GF NYLON 1.5IN
Supplier:  Cole-Parmer 5011882
Encompass_Preferred
Key FeaturesMeet US and EU standards for use in pharmaceutical process applicationsASME-BPE, FDA, and USP Class VI compliantLightweight and reusableRust and corrosion resistantCompatible with Tri-Clamp® fittingsSterilize by autoclave or gamma irradiationIdeal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your systemFree technical application support available to determine which connector type is needed for your application1-1/2 in. clamp sizeManufacturer defect warranty
Catalog No. NC3760921