Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Tee, Sanitary Clamp Union, 1 1/2"" Clamp x 1 1/2"" Clamp x 1 1/2"" Clamp
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3180663/DEL
Encompass
Tri-Clamp standard tee adapters allow for liquid flow to be dividing into two or combined into one flow path. Though both materials may be autoclaved, PVDF tolerates high temperatures better than PP does and will perform more consistently after repeated autoclaving. Clamps and gaskets required for a complete system are sold separately.
Catalog No. NC0016398