Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) SQCC221212M Quick-disconnect insert, stainless steel, 3/4"" sanitary connector
Catalog No. NC1626939
Encompass
These couplings allow you to integrate disposables with your existing stainless steel processing equipment. Both couplings and inserts are available with standard sanitary terminations to attach to your system with a sanitary gasket and tri-clover clamps. Choose from stainless steel or polysulfone inserts to mate to polysulfone bodies. All models feature USP Class VI platinum-cured silicone gaskets. Mix and match these bodies and inserts with the polysulfone quick-disconnect coupling elements on page 509 for greater flexibility in your process system.
Catalog No. NC1626939 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3160430
These couplings allow you to integrate disposables with your existing stainless steel processing equipment. Both couplings and inserts are available with standard sanitary terminations to attach to your system with a sanitary gasket and tri-clover clamps. Choose from stainless steel or polysulfone inserts to mate to polysulfone bodies. All models feature USP Class VI platinum-cured silicone gaskets. Mix and match these bodies and inserts with the polysulfone quick-disconnect coupling elements on page 509 for greater flexibility in your process system.