Learn More
Hach Company Sanitary Dig. Conduc. Sen 0.05
Catalog No. 5000208129 Shop All Hach Company Products
Encompass
Digital Sanitary (CIP) Contacting Conductivity Sensor for Low Conductivity-k 0 05 Cell ConstantNominal Cell Constant (K) 0 05 Sanitary (Cip) 1-1/2 Inch Flange 6M (20 ft) Analog Cable 316 Stainless Steel Electrode Includes Digital Gateway And 1M (3 3 ft) Digital Extension Cable
Catalog No. 50-002-08129 Supplier Hach Company Supplier No. D3455A6
Digital Sanitary (CIP) Contacting Conductivity Sensor for Low Conductivity-k 0 05 Cell ConstantNominal Cell Constant (K) 0 05 Sanitary (Cip) 1-1/2 Inch Flange 6M (20 ft) Analog Cable 316 Stainless Steel Electrode Includes Digital Gateway And 1M (3 3 ft) Digital Extension Cable