Learn More
NORDSON MEDICAL (NH) SANITARY FITTING MAXI FLANGE
Catalog No. NC3622004
Encompass_Preferred
Sanitary Fitting, Maxi Flange to 600 Series Barb, 3/8" (9.5 mm) Tubing ID, Natural Kynar PVDF 100 PIECES MINIMUM ORDER
Catalog No. NC3622004 Supplier NORDSON MEDICAL (NH) Supplier No. SFMX670J2A
Sanitary Fitting, Maxi Flange to 600 Series Barb, 3/8" (9.5 mm) Tubing ID, Natural Kynar PVDF 100 PIECES MINIMUM ORDER