Learn More
Hach Company Sanitary Mounting Kit, 3700 Series and pHD Sensors
Catalog No. 501998047 Shop All Hach Company Products
Encompass
316 Stainless Steel Sanitary 2-Inch Tee and Heavy Duty Clamp. For use with the sanitary style 3700-series inductive/electrodeless conductivity sensors.
Catalog No. 50-199-8047 Supplier Hach Company Supplier No. MH018S8SZ
316 Stainless Steel Sanitary 2-Inch Tee and Heavy Duty Clamp. For use with the sanitary style 3700-series inductive/electrodeless conductivity sensors.