Learn More
MSC American Specialties Sanitary Napkin Receptacle, Surface Mounted, Stainless Steel, 1.5 Gallon
Catalog No. 502837754
Encompass
This sanitary napkin receptacle is designed for surface wall mounting and offers a discreet and efficient solution for waste disposal in commercial and public restrooms. Constructed from durable stainless steel, it features a 1.5-gallon capacity and a self-closing door, ensuring hygiene and ease of maintenance.
  • Receptacle is removable for servicing.
  • Locks to the back of the cabinet using stainless steel clips.
  • Features a self-closing door mounted with a heavy-duty, full-length piano hinge.
  • Fabricated from alloy 18-8 stainless steel, type 304, 22 gauge.
  • Exposed surfaces have a No. 4 satin finish.
  • Welded construction with no exposed fasteners or visible spot-welded seams.
  • Waste container is captured internally by a full-width Z-retainer.
  • Safety-edged finger grip provides convenient grip for service.
Catalog No. 50-283-7754 Supplier MSC Supplier No. 62916895
This sanitary napkin receptacle is designed for surface wall mounting and offers a discreet and efficient solution for waste disposal in commercial and public restrooms. Constructed from durable stainless steel, it features a 1.5-gallon capacity and a self-closing door, ensuring hygiene and ease of maintenance.
  • Receptacle is removable for servicing.
  • Locks to the back of the cabinet using stainless steel clips.
  • Features a self-closing door mounted with a heavy-duty, full-length piano hinge.
  • Fabricated from alloy 18-8 stainless steel, type 304, 22 gauge.
  • Exposed surfaces have a No. 4 satin finish.
  • Welded construction with no exposed fasteners or visible spot-welded seams.
  • Waste container is captured internally by a full-width Z-retainer.
  • Safety-edged finger grip provides convenient grip for service.