Learn More
AVANTOR MASTERFLEX (VWR LLC) Sanitary Plastic Ball Valve, 2-Way, 3/4"" Mini Tri-Clamp, Polypropylene Body, EPDM Seals
Catalog No. NC1675731
Encompass
These industry standard valves feature PTFE bearings and seats to virtually eliminate friction and minimize wear. Fully concentric polished ball further ensures leak-free performance. Available in polyproplyene (PP) or PVDF body material with EPDM seals.
Catalog No. NC1675731 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3053201
These industry standard valves feature PTFE bearings and seats to virtually eliminate friction and minimize wear. Fully concentric polished ball further ensures leak-free performance. Available in polyproplyene (PP) or PVDF body material with EPDM seals.