Learn More
Cole-Parmer Wika M932.25 2.5" Sanitary Pressure Gauge, 0 to 30 psi, 3/4 Tri-Clamp
Catalog No. NC9642303 Shop All Cole-Parmer Products
Encompass_Preferred
Gauges all-welded assembly contains an external flush diaphragm and is ideal for applications in the food, beverage, pharmaceutical, and biotechnology industries. Gauges are constructed of 316 stainless steel and have a 3/4" Tri-Clamp process connection welded to the gauge. Assembly meets the criteria set forth by 3A standards. Gauge case and ring are 304 stainless steel and has a polycarbonate window.
Catalog No. NC9642303 Supplier Cole-Parmer Supplier No. 6805672
Gauges all-welded assembly contains an external flush diaphragm and is ideal for applications in the food, beverage, pharmaceutical, and biotechnology industries. Gauges are constructed of 316 stainless steel and have a 3/4" Tri-Clamp process connection welded to the gauge. Assembly meets the criteria set forth by 3A standards. Gauge case and ring are 304 stainless steel and has a polycarbonate window.