Learn More
Accuflow Systems SANITECH TUBING C250-2 50FT
Catalog No. NC9746504
Encompass_Preferred
Features/Benefits: • Ultra-pure biopharmaceutical-grade silicone • Unsurpassed surface smoothness • Ultra-flexible • Autoclavable and gamma stable • Temperature range from -80°F (-62°C) to 500°F (260°C) • Imparts no taste or odors • Laser-marked part numbers and lot codes (1/16'' ID and greater) • Color striping available • Available in bulk or custom assemblies and multiple length coils • Meets USP Class VI, European Pharmacopoeia 3.1.9 and FDA criteria*
Catalog No. NC9746504 Supplier Accuflow Systems Supplier No. STHTC2502F
Features/Benefits: • Ultra-pure biopharmaceutical-grade silicone • Unsurpassed surface smoothness • Ultra-flexible • Autoclavable and gamma stable • Temperature range from -80°F (-62°C) to 500°F (260°C) • Imparts no taste or odors • Laser-marked part numbers and lot codes (1/16'' ID and greater) • Color striping available • Available in bulk or custom assemblies and multiple length coils • Meets USP Class VI, European Pharmacopoeia 3.1.9 and FDA criteria*