Learn More
Essendant Beer Clean Last Rinse Glass Sanitizer, Powder, .25oz Packet, 100/Carton
Catalog No. 502004439
Encompass_Preferred
Choose a tried-and-true glassware sanitzer that's trusted by and used in breweries throughout North America. Powdered compound contains trichloromelamine (TCM), ensuring effective sanitization of all glassware. Laboratory tested¿kills E. coli and staph. Formulated for use in cold water.
Catalog No. 50-200-4439 Supplier Essendant Supplier No. DVO90223
Choose a tried-and-true glassware sanitzer that's trusted by and used in breweries throughout North America. Powdered compound contains trichloromelamine (TCM), ensuring effective sanitization of all glassware. Laboratory tested¿kills E. coli and staph. Formulated for use in cold water.