Learn More
Med Vet International McKesson Foaming Hand Sanitizer with Aloe 1000 mL, Dispenser Refill Bag, 53-29036-1000, Each
Catalog No. 503045396
Encompass_Preferred
Hand Sanitizer with Aloe McKesson 1,000 mL Ethyl Alcohol Foaming Dispenser Refill Bag Refill for McKesson Pump Foam Dispenser (MFR # 53-FOAM) and McKesson Automatic Foam Dispenser (MFR # 53-FOAM-AUTO) Contains Vitamin E, moisturizers and emollients to help prevent drying of the skin with frequent use A no-rinse, moisturizing hand gel helps decrease bacteria on skin Helps protect against cross contamination Convenient and ready to use; no water or towels needed Made with aloe to help keep skin soft
Catalog No. 50-304-5396 Supplier Med Vet International Supplier No. 957986EA
Hand Sanitizer with Aloe McKesson 1,000 mL Ethyl Alcohol Foaming Dispenser Refill Bag Refill for McKesson Pump Foam Dispenser (MFR # 53-FOAM) and McKesson Automatic Foam Dispenser (MFR # 53-FOAM-AUTO) Contains Vitamin E, moisturizers and emollients to help prevent drying of the skin with frequent use A no-rinse, moisturizing hand gel helps decrease bacteria on skin Helps protect against cross contamination Convenient and ready to use; no water or towels needed Made with aloe to help keep skin soft