Learn More
Med Vet International McKesson Premium Hand Sanitizer With Aloe, Ethyl Alcohol Gel, 2oz, 53-27039-2, 48/Case
Catalog No. 503045334
Encompass_Preferred
Hand Sanitizer with Aloe McKesson Premium 2 oz. Ethyl Alcohol Gel Bottle McKesson premium hand sanitizer with aloe kills over 99 percent of bacteria with its ethyl alcohol solution It soothes the skin without leaving behind residue Ideal for use when handwashing with soap and water isn't available This ethyl alcohol hand sanitizer gel contains vitamin E and emollients to moisturize the skin and prevent it from drying out The container is easy to store on a counter or shelf
Catalog No. 50-304-5334 Supplier Med Vet International Supplier No. 937912CS
Hand Sanitizer with Aloe McKesson Premium 2 oz. Ethyl Alcohol Gel Bottle McKesson premium hand sanitizer with aloe kills over 99 percent of bacteria with its ethyl alcohol solution It soothes the skin without leaving behind residue Ideal for use when handwashing with soap and water isn't available This ethyl alcohol hand sanitizer gel contains vitamin E and emollients to moisturize the skin and prevent it from drying out The container is easy to store on a counter or shelf