Learn More
Med Vet International McKesson Premium Hand Sanitizer with Aloe, 8 oz., 53-27033-8C, Each
Catalog No. 503043268
Encompass_Preferred
Hand Sanitizer with Aloe McKesson Premium 8 oz. Ethyl Alcohol Gel Bottle Convenient and ready to use with no water or towels needed No-rinse, moisturizing hand gel with Vitamin E Includes aloe to keep skin soft without leaving a sticky residue 70% Ethanol formula designed to kill more than 99% of most bacteria on skin to help protect against cross-contamination Slick gel solution flows easily to access all areas of the hand Spring water fragrance leaves a fresh scent
Catalog No. 50-304-3268 Supplier Med Vet International Supplier No. 1162514EA
Hand Sanitizer with Aloe McKesson Premium 8 oz. Ethyl Alcohol Gel Bottle Convenient and ready to use with no water or towels needed No-rinse, moisturizing hand gel with Vitamin E Includes aloe to keep skin soft without leaving a sticky residue 70% Ethanol formula designed to kill more than 99% of most bacteria on skin to help protect against cross-contamination Slick gel solution flows easily to access all areas of the hand Spring water fragrance leaves a fresh scent