Learn More
Med Vet International McKesson Premium Hand Sanitizer with Aloe, 8 oz., Gel, Pump Bottle, 53-27033-8, Each
Catalog No. 503045328
Encompass_Preferred
Hand Sanitizer with Aloe McKesson Premium 8 oz. Ethyl Alcohol Gel Pump Bottle The McKesson premium hand sanitizer with aloe kills over 99 percent of bacteria with its ethyl alcohol solution It soothes the skin without leaving behind residue Ideal for use when handwashing with soap and water isn't available This ethyl alcohol hand sanitizer gel contains vitamin E and emollients to moisturize the skin and prevent it from drying out The container is easy to store on a counter or shelf
Catalog No. 50-304-5328 Supplier Med Vet International Supplier No. 937915EA
Hand Sanitizer with Aloe McKesson Premium 8 oz. Ethyl Alcohol Gel Pump Bottle The McKesson premium hand sanitizer with aloe kills over 99 percent of bacteria with its ethyl alcohol solution It soothes the skin without leaving behind residue Ideal for use when handwashing with soap and water isn't available This ethyl alcohol hand sanitizer gel contains vitamin E and emollients to moisturize the skin and prevent it from drying out The container is easy to store on a counter or shelf