Learn More
Med Vet International Hand Sanitizer Purell Healthcare Advanced 1,200 mL Ethyl Alcohol Foaming Dispenser Refill Bottle, 7753-02, 2/Case
Catalog No. 503044737
Encompass_Preferred
1200 mL, scented Advanced foam hand sanitizer formulation designed for healthcare environments Kills more than 99.99% of most common germs that may cause illness in a healthcare setting , including MRSA and VRE Enhanced with a nourishing blend of four skin conditioners, clinically proven to maintain skin health Chlorhexidine Gluconate (CHG) compatible formulation Compatible with nitrile, vinyl, and latex gloves Refill for Purell ES8 Touch-Free Hand Sanitizer Dispensers
Catalog No. 50-304-4737 Supplier Med Vet International Supplier No. 1087441CS
1200 mL, scented Advanced foam hand sanitizer formulation designed for healthcare environments Kills more than 99.99% of most common germs that may cause illness in a healthcare setting , including MRSA and VRE Enhanced with a nourishing blend of four skin conditioners, clinically proven to maintain skin health Chlorhexidine Gluconate (CHG) compatible formulation Compatible with nitrile, vinyl, and latex gloves Refill for Purell ES8 Touch-Free Hand Sanitizer Dispensers