Learn More
Med Vet International Hand Sanitizer Purell Advanced 1,200 mL Ethyl Alcohol Gel Dispenser Refill Bottle, 1903-02, 2/Case
Catalog No. 503044121
Encompass_Preferred
The Purell Advanced Hand Sanitizer Gel contains 70 percent ethyl alcohol that kills 99.99 percent of germs and bacteria It meets ECOLOGO hand sanitizer standard It meets the USDA BioPreferred Program for biobased products The SMART-FLEX™ design in this 1,200 mL refill bottle uses less material It is green certified and has a removable pump Hygienic SANITARY SEALED™ refills retain bottle shape using Controlled Collapse™ technology
Catalog No. 50-304-4121 Supplier Med Vet International Supplier No. 796466CS
The Purell Advanced Hand Sanitizer Gel contains 70 percent ethyl alcohol that kills 99.99 percent of germs and bacteria It meets ECOLOGO hand sanitizer standard It meets the USDA BioPreferred Program for biobased products The SMART-FLEX™ design in this 1,200 mL refill bottle uses less material It is green certified and has a removable pump Hygienic SANITARY SEALED™ refills retain bottle shape using Controlled Collapse™ technology