Learn More
Med Vet International Hand Sanitizer Purell Advanced 4.25 oz. Ethyl Alcohol Gel Bottle, 9651-24, Each
Catalog No. 503043644
Encompass_Preferred
Purell hand sanitizer in a bottle contains four skin conditioners It keeps your hands soft and soothed Purell advanced hand sanitizer gel contains 70 percent ethyl alcohol It kills 99.99 percent of common germs It's easy to use and doesn't require water or towels It takes only 15 seconds to become effective in the fight against contracting germs It comes in a convenient 4 oz bottle for quick dispensing
Catalog No. 50-304-3644 Supplier Med Vet International Supplier No. 281327EA
Purell hand sanitizer in a bottle contains four skin conditioners It keeps your hands soft and soothed Purell advanced hand sanitizer gel contains 70 percent ethyl alcohol It kills 99.99 percent of common germs It's easy to use and doesn't require water or towels It takes only 15 seconds to become effective in the fight against contracting germs It comes in a convenient 4 oz bottle for quick dispensing