Learn More
Med Vet International Hand Sanitizer Purell Advanced 12 oz. Ethyl Alcohol Gel Pump Bottle, 3659-12, 12/Case
Catalog No. 503044315
Encompass_Preferred
The gel kills 99.99 percent of common germs that cause illness The formula in this 70 percent strength ethyl alcohol hand sanitizer gel leaves hands refreshed Four skin conditioners and a fruit scent help improve skin health and freshness The gel is free of dye, triclosan, parabens, phthalates or harsh preservatives Purell Advanced hand sanitizer refreshing gel evaporates quickly It is available in a handy 12 ounce table top pump bottle
Catalog No. 50-304-4315 Supplier Med Vet International Supplier No. 714107CS
 Restricted Item
Add to Cart
Add to Cart
The gel kills 99.99 percent of common germs that cause illness The formula in this 70 percent strength ethyl alcohol hand sanitizer gel leaves hands refreshed Four skin conditioners and a fruit scent help improve skin health and freshness The gel is free of dye, triclosan, parabens, phthalates or harsh preservatives Purell Advanced hand sanitizer refreshing gel evaporates quickly It is available in a handy 12 ounce table top pump bottle