Learn More
Med Vet International PediFix ShoeZap 15 Minute UV Shoe Sanitizer, P3410/10, Each
Catalog No. 503042955
Encompass_Preferred
ShoeZap UV Shoe Sanitizer Kills fungus and bacteria that cause athlete's foot, fungal nails, foot and shoe odor, and diabetic infections Keeps shoes clean and hygienic without toxic chemical sprays Clinically proven technology Easy, safe and effective 15-minute automatic-off time cycle
Catalog No. 50-304-2955 Supplier Med Vet International Supplier No. 1112157EA
ShoeZap UV Shoe Sanitizer Kills fungus and bacteria that cause athlete's foot, fungal nails, foot and shoe odor, and diabetic infections Keeps shoes clean and hygienic without toxic chemical sprays Clinically proven technology Easy, safe and effective 15-minute automatic-off time cycle