Learn More
Essendant Foodservice Surface Sanitizer, Fragrance Free, 32 oz Capped Bottle with Spray Trigger Included in Carton, 6/Carton
Catalog No. 2790186
Encompass_Preferred
Formulated for convenience and ease-of-use, Foodservice Surface Sanitizer Spray provides powerful germ-kill on the surfaces people touch most ¿ yet is gentle enough to use on a child¿s highchair with no rinse required. Patented fragrance-free formulation sanitizes without any harsh chemicals. It eliminates 99.9 percent of viruses and bacteria on surfaces ¿ including norovirus, E. coli, Salmonella, and human coronavirus in 30 seconds. With the EPA¿s lowest allowable toxicity rating, it delivers the overall fastest disinfection time of any Design for the Environment (DfE) product.
Catalog No. 27-901-86 Supplier Essendant Supplier No. GOJ334106CT
Formulated for convenience and ease-of-use, Foodservice Surface Sanitizer Spray provides powerful germ-kill on the surfaces people touch most ¿ yet is gentle enough to use on a child¿s highchair with no rinse required. Patented fragrance-free formulation sanitizes without any harsh chemicals. It eliminates 99.9 percent of viruses and bacteria on surfaces ¿ including norovirus, E. coli, Salmonella, and human coronavirus in 30 seconds. With the EPA¿s lowest allowable toxicity rating, it delivers the overall fastest disinfection time of any Design for the Environment (DfE) product.