Learn More
Med Vet International Hand Sanitizer Purell Advanced 1,200 mL Ethyl Alcohol Foaming Dispenser Refill Bottle, 1905-02, Each
Catalog No. 503043435
Encompass_Preferred
The Purell hand sanitizer 1200 milliliter refill option features a 70 percent ethyl alcohol formulation for high efficacy The foaming consistency sanitizes while remaining gentle on your skin The container uses the least amount of material possible and a removable pump for an easily recyclable product A removable dispenser pump makes refills more hygienic Purell Advanced hand sanitizer foam uses GOJO Sanitary Sealed™ technology to prevent foreign contaminants in your product It has a light fruit scent
Catalog No. 50-304-3435 Supplier Med Vet International Supplier No. 796445EA
The Purell hand sanitizer 1200 milliliter refill option features a 70 percent ethyl alcohol formulation for high efficacy The foaming consistency sanitizes while remaining gentle on your skin The container uses the least amount of material possible and a removable pump for an easily recyclable product A removable dispenser pump makes refills more hygienic Purell Advanced hand sanitizer foam uses GOJO Sanitary Sealed™ technology to prevent foreign contaminants in your product It has a light fruit scent