Learn More
Med Vet International Hand Sanitizer Isagel 21 oz. Ethyl Alcohol Gel Pump Bottle, 1645, 12/Case
Catalog No. 503044250
Encompass_Preferred
The Isagel hand sanitizer eliminates 99.99 percent of illness-causing microorganisms It has a non-rinse formula and is ideal in situations without access to soap and water It has moisturizers that prevent the skin from drying after cleansing The instant hand sanitizing gel made with ethyl alcohol has 60 percent strength It is compatible with latex gloves The skin cleanser works within 15 seconds It is packaged in a 21 oz pump bottle and has an alcohol scent
Catalog No. 50-304-4250 Supplier Med Vet International Supplier No. 548522CS
The Isagel hand sanitizer eliminates 99.99 percent of illness-causing microorganisms It has a non-rinse formula and is ideal in situations without access to soap and water It has moisturizers that prevent the skin from drying after cleansing The instant hand sanitizing gel made with ethyl alcohol has 60 percent strength It is compatible with latex gloves The skin cleanser works within 15 seconds It is packaged in a 21 oz pump bottle and has an alcohol scent