Learn More
Electron Microscopy Sciences SaniZide™ Germicidal Solution 32 OZ
SDP
Catalog No. 50930403
Encompass_Preferred
SaniZide Plus is a ready to use, hospital grade, hard surface disinfectant deodorizer is quatrernary ammonium, alcohol free formulation is non-flammable making it safe to ship, as well as non-corrosive. Choosing SaniZide Plus™ helps you to comply with the OSHA Bloodborne Pathogens Standard, which requires the use of an "appropriate disinfectant".
Safe for useon: glass, porcelain, ceramic, metal, polyethylene, polypropylene, vinyl, polyester, rubber, bakelite, and many more.
This item is not returnable. View return policy
Catalog No. 50-930-403 Supplier Electron Microscopy Sciences Supplier No. 7058032
SaniZide Plus is a ready to use, hospital grade, hard surface disinfectant deodorizer is quatrernary ammonium, alcohol free formulation is non-flammable making it safe to ship, as well as non-corrosive. Choosing SaniZide Plus™ helps you to comply with the OSHA Bloodborne Pathogens Standard, which requires the use of an "appropriate disinfectant".
Safe for useon: glass, porcelain, ceramic, metal, polyethylene, polypropylene, vinyl, polyester, rubber, bakelite, and many more.