Learn More
Selleck Chemical LLC SANT-1 10mg 304909-07-7
SDP
Supplier:  Selleck Chemical LLC S709210MG
Encompass_Preferred
SANT-1 directly binds to Smoothened (Smo) receptor with Kd of 1.2 nM and inhibits Smo agonist effects with IC50 of 20 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4390