Learn More
Medchemexpress LLC HY-100224 10mg Medchemexpress, SANT-1 CAS:304909-07-7 Purity:>98%
SDP
Catalog No. 501967716
Encompass_Preferred
Medchemexpress, HY-100224 10mg SANT-1 CAS:304909-07-7 SANT-1 is a potent Smo antagonist, inhibits Hedgehog signaling, with IC50s of 20 nM and 30 nM in Shh-LIGHT2 and SmoA1-LIGHT2 assay, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7716 Supplier Medchemexpress LLC Supplier No. HY10022410MG
Medchemexpress, HY-100224 10mg SANT-1 CAS:304909-07-7 SANT-1 is a potent Smo antagonist, inhibits Hedgehog signaling, with IC50s of 20 nM and 30 nM in Shh-LIGHT2 and SmoA1-LIGHT2 assay, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.