Learn More
Enzo Life Sciences SANT-1 (1mg). CAS: 304909-07-7
SDP
Catalog No. 502013404
Encompass_Preferred
Equipotently inhibits the activities of both wild-type and oncogenic Smoothened. Alternative name: Shh signaling antagonist V, (E)-N-(4-benzylpiperazin-1-yl)-1-(3,5-dimethyl-1-phenylpyrazol-4-yl)methanimine. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or ethanol. Long Term Storage: -80°C.
Catalog No. 50-201-3404 Supplier Enzo Life Sciences Supplier No. ENZCHM1200001
Equipotently inhibits the activities of both wild-type and oncogenic Smoothened. Alternative name: Shh signaling antagonist V, (E)-N-(4-benzylpiperazin-1-yl)-1-(3,5-dimethyl-1-phenylpyrazol-4-yl)methanimine. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or ethanol. Long Term Storage: -80°C.